Assay Details
| Gene Symbol: LOC4341997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787519.1 | XP_015643005.1 | 5 - 6 | 725 | 77 | |
| XM_015787521.1 | XP_015643007.1 | 4 - 5 | 561 | 77 | ||
| GenBank mRNA | AK065784.1 | - | 5 - 6 | 617 | 77 | |
| AK101836.1 | - | 5 - 6 | 716 | 77 | ||
| AK119551.1 | - | 5 - 6 | 609 | 77 | ||
More Information
Set Membership: |
|

LOC4341997
Genomic Map