Assay Details
| Gene Symbol: LOC4341997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787519.1 | XP_015643005.1 | 7 - 8 | 869 | 81 | |
| XM_015787521.1 | XP_015643007.1 | 6 - 7 | 705 | 81 | ||
| GenBank mRNA | AK065784.1 | - | 7 - 8 | 761 | 81 | |
| AK101836.1 | - | 7 - 8 | 860 | 81 | ||
| AK119551.1 | - | 7 - 8 | 753 | 81 | ||
More Information
Set Membership: |
|

LOC4341997
Genomic Map