Assay Details
| Gene Symbol: LOC4342019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787586.2 | XP_015643072.1 | 5 - 6 | 584 | 65 | |
| GenBank mRNA | AK103849.1 | - | 4 - 5 | 494 | 65 | |
More Information
Set Membership: |
|

LOC4342019
Genomic Map