Assay Details
| Gene Symbol: LOC4342019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787586.2 | XP_015643072.1 | 11 - 12 | 1347 | 75 | |
| GenBank mRNA | AK068246.1 | - | 2 - 3 | 3009 | 75 | |
| AK103849.1 | - | 8 - 9 | 2093 | 75 | ||
More Information
Set Membership: |
|

LOC4342019
Genomic Map