Assay Details
| Gene Symbol: LOC4342043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787723.2 | XP_015643209.1 | 12 - 13 | 1563 | 95 | |
| GenBank mRNA | AK058857.1 | - | 1 - 2 | 142 | 95 | |
| AK070744.1 | - | 12 - 13 | 1572 | 95 | ||
More Information
Set Membership: |
|

LOC4342043
Genomic Map