Assay Details
| Gene Symbol: LOC4342043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787723.2 | XP_015643209.1 | 2 - 3 | 709 | 101 | |
| GenBank mRNA | AK070744.1 | - | 2 - 3 | 718 | 101 | |
More Information
Set Membership: |
|

LOC4342043
Genomic Map