Assay Details
| Gene Symbol: LOC4342043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787723.2 | XP_015643209.1 | 6 - 7 | 1061 | 76 | |
| GenBank mRNA | AK070744.1 | - | 6 - 7 | 1070 | 76 | |
More Information
Set Membership: |
|

LOC4342043
Genomic Map