Assay Details
| Gene Symbol: LOC4342163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791033.2 | XP_015646519.1 | 1 - 2 | 555 | 75 | |
| GenBank mRNA | AK066040.1 | - | 1 - 2 | 498 | 75 | |
| AK109623.1 | - | 1 - 2 | 498 | 75 | ||
More Information
Set Membership: |
|

LOC4342163
Genomic Map