Assay Details
| Gene Symbol: LOC4342163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791033.2 | XP_015646519.1 | 9 - 10 | 1175 | 68 | |
| GenBank mRNA | AK061192.1 | - | 4 - 5 | 308 | 68 | |
| AK066040.1 | - | 9 - 10 | 1118 | 68 | ||
More Information
Set Membership: |
|

LOC4342163
Genomic Map