Assay Details
| Gene Symbol: LOC4342403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791887.2 | XP_015647373.1 | 30 - 31 | 4258 | 107 | |
| XM_015791888.2 | XP_015647374.1 | 30 - 31 | 4192 | 107 | ||
| GenBank mRNA | AK062246.1 | - | 2 - 3 | 119 | 107 | |
More Information
Set Membership: |
|

LOC4342403
Genomic Map