Assay Details
| Gene Symbol: LOC4342403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791887.2 | XP_015647373.1 | 32 - 33 | 4586 | 91 | |
| XM_015791888.2 | XP_015647374.1 | 32 - 33 | 4520 | 91 | ||
| GenBank mRNA | AK062246.1 | - | 4 - 5 | 447 | 91 | |
More Information
Set Membership: |
|

LOC4342403
Genomic Map