Assay Details
Gene Symbol: LOC4342403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791887.2 | XP_015647373.1 | 36 - 37 | 4963 | 107 | |
XM_015791888.2 | XP_015647374.1 | 36 - 37 | 4897 | 107 | ||
GenBank mRNA | AK062246.1 | - | 8 - 9 | 824 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
