Assay Details
Gene Symbol: LOC4342403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791887.2 | XP_015647373.1 | 37 - 38 | 5096 | 75 | |
XM_015791888.2 | XP_015647374.1 | 37 - 38 | 5030 | 75 | ||
GenBank mRNA | AK062246.1 | - | 9 - 10 | 957 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
