Assay Details
| Gene Symbol: LOC4342575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789522.2 | XP_015645008.1 | 13 - 14 | 4707 | 76 | |
| XM_015789523.2 | XP_015645009.1 | 13 - 14 | 4745 | 76 | ||
| GenBank mRNA | AK111461.1 | - | 7 - 8 | 1196 | 76 | |
More Information
Set Membership: |
|

LOC4342575
Genomic Map