Assay Details
| Gene Symbol: LOC4342585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791254.2 | XP_015646740.1 | 1 - 2 | 247 | 67 | |
| GenBank mRNA | AK072273.1 | - | 1 - 2 | 199 | 67 | |
| AK100930.1 | - | 1 - 2 | 209 | 67 | ||
More Information
Set Membership: |
|

LOC4342585
Genomic Map