Assay Details
| Gene Symbol: LOC4342585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791254.2 | XP_015646740.1 | 2 - 3 | 287 | 62 | |
| GenBank mRNA | AK072273.1 | - | 2 - 3 | 239 | 62 | |
| AK100930.1 | - | 2 - 3 | 249 | 62 | ||
More Information
Set Membership: |
|

LOC4342585
Genomic Map