Assay Details
| Gene Symbol: LOC4342652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK069093.1 | - | 1 - 2 | 470 | 82 | |
| AK073823.1 | - | 1 - 2 | 470 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4342652| Gene Symbol: LOC4342652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK069093.1 | - | 1 - 2 | 470 | 82 | |
| AK073823.1 | - | 1 - 2 | 470 | 82 | ||
Set Membership: |
|