Assay Details
| Gene Symbol: LOC4342946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789915.1 | XP_015645401.1 | 1 | 598 | 107 | |
| GenBank mRNA | AK062503.1 | - | 1 | 552 | 107 | |
More Information
Set Membership: |
|

LOC4342946
Genomic Map