Assay Details
| Gene Symbol: LOC4342990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789938.2 | XP_015645424.1 | 17 - 18 | 1401 | 87 | |
| GenBank mRNA | AK064087.1 | - | 13 - 14 | 971 | 87 | |
| AK101812.1 | - | 17 - 18 | 1341 | 87 | ||
More Information
Set Membership: |
|

LOC4342990
Genomic Map