Assay Details
| Gene Symbol: LOC4342990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789938.2 | XP_015645424.1 | 18 - 19 | 1516 | 81 | |
| GenBank mRNA | AK064087.1 | - | 14 - 15 | 1086 | 81 | |
| AK101812.1 | - | 18 - 19 | 1456 | 81 | ||
More Information
Set Membership: |
|

LOC4342990
Genomic Map