Assay Details
| Gene Symbol: LOC4342990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789938.2 | XP_015645424.1 | 4 - 5 | 554 | 76 | |
| GenBank mRNA | AK101812.1 | - | 4 - 5 | 494 | 76 | |
More Information
Set Membership: |
|

LOC4342990
Genomic Map