Assay Details
| Gene Symbol: LOC4343108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791774.1 | XP_015647260.1 | 4 - 5 | 903 | 124 | |
| GenBank mRNA | AK074000.1 | - | 4 - 5 | 909 | 124 | |
| AK107130.1 | - | 3 - 4 | 873 | 203 | ||
More Information
Set Membership: |
|

LOC4343108
Genomic Map