Assay Details
| Gene Symbol: LOC4343122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791342.2 | XP_015646828.1 | 2 - 3 | 882 | 107 | |
| GenBank mRNA | AK072519.1 | - | 2 - 3 | 867 | 107 | |
| AK103938.1 | - | 2 - 3 | 866 | 107 | ||
| AK103970.1 | - | 2 - 3 | 864 | 107 | ||
| AK119656.1 | - | 2 - 3 | 864 | 107 | ||
| KJ909972.1 | - | 2 - 3 | 849 | 107 | ||
More Information
Set Membership: |
|

LOC4343122
Genomic Map