Assay Details
Gene Symbol: LOC4343132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791831.2 | XP_015647317.1 | 3 - 4 | 789 | 144 | |
GenBank mRNA | AK287638.1 | - | 3 - 4 | 759 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
