Assay Details
Gene Symbol: LOC4343241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791778.1 | XP_015647264.1 | 9 - 10 | 1281 | 113 | |
GenBank mRNA | AK060926.1 | - | 4 - 5 | 440 | 113 | |
AK072721.1 | - | 9 - 10 | 1281 | 113 | ||
HQ324821.1 | - | 9 - 10 | 1271 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
