Assay Details
| Gene Symbol: LOC4343342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789784.1 | XP_015645270.1 | 6 - 7 | 966 | 91 | |
| GenBank mRNA | AK064439.1 | - | 2 - 3 | 168 | 91 | |
More Information
Set Membership: |
|

LOC4343342
Genomic Map