Assay Details
| Gene Symbol: LOC4343342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789784.1 | XP_015645270.1 | 9 - 10 | 1229 | 140 | |
| GenBank mRNA | AK064439.1 | - | 5 - 6 | 431 | 140 | |
More Information
Set Membership: |
|

LOC4343342
Genomic Map