Assay Details
| Gene Symbol: LOC4343342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789784.1 | XP_015645270.1 | 11 - 12 | 1448 | 63 | |
| GenBank mRNA | AK064439.1 | - | 7 - 8 | 650 | 63 | |
More Information
Set Membership: |
|

LOC4343342
Genomic Map