Assay Details
| Gene Symbol: LOC4343357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792005.2 | XP_015647491.1 | 1 - 2 | 265 | 113 | |
| XM_015792006.2 | XP_015647492.1 | 1 - 2 | 266 | 113 | ||
| GenBank mRNA | AK070461.1 | - | 1 - 2 | 262 | 113 | |
More Information
Set Membership: |
|

LOC4343357
Genomic Map