Assay Details
| Gene Symbol: LOC4343745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791564.2 | XP_015647050.1 | 6 - 7 | 1419 | 139 | |
| GenBank mRNA | AK240739.1 | - | 6 - 7 | 1104 | 139 | |
More Information
Set Membership: |
|

LOC4343745
Genomic Map