Assay Details
Gene Symbol: LOC4343748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790074.2 | XP_015645560.1 | 1 - 2 | 379 | 62 | |
GenBank mRNA | AK071404.1 | - | 1 - 2 | 346 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
