Assay Details
Gene Symbol: LOC4343748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790074.2 | XP_015645560.1 | 3 - 4 | 763 | 87 | |
GenBank mRNA | AK071404.1 | - | 3 - 4 | 730 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
