Assay Details
| Gene Symbol: LOC4343846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791445.2 | XP_015646931.1 | 1 - 2 | 796 | 77 | |
| GenBank mRNA | AK064312.1 | - | 1 - 2 | 783 | 77 | |
| AK069085.1 | - | 1 - 2 | 785 | 77 | ||
More Information
Set Membership: |
|

LOC4343846
Genomic Map