Assay Details
| Gene Symbol: LOC4343927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791470.2 | XP_015646956.1 | 6 - 7 | 1153 | 65 | |
| XM_015791471.2 | XP_015646957.1 | 5 - 6 | 651 | 65 | ||
| GenBank mRNA | AK064123.1 | - | 7 - 8 | 1061 | 65 | |
More Information
Set Membership: |
|

LOC4343927
Genomic Map