Assay Details
| Gene Symbol: LOC4343937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791131.2 | XP_015646617.1 | 1 | 1664 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4343937| Gene Symbol: LOC4343937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791131.2 | XP_015646617.1 | 1 | 1664 | 137 | |
Set Membership: |
|