Assay Details
| Gene Symbol: LOC4343938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK059824.1 | - | 2 - 3 | 186 | 110 | |
| AK071567.1 | - | 2 - 3 | 242 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4343938| Gene Symbol: LOC4343938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK059824.1 | - | 2 - 3 | 186 | 110 | |
| AK071567.1 | - | 2 - 3 | 242 | 110 | ||
Set Membership: |
|