Assay Details
| Gene Symbol: LOC4343938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK059824.1 | - | 4 - 5 | 562 | 89 | |
| AK071567.1 | - | 4 - 5 | 618 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4343938| Gene Symbol: LOC4343938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK059824.1 | - | 4 - 5 | 562 | 89 | |
| AK071567.1 | - | 4 - 5 | 618 | 89 | ||
Set Membership: |
|