Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 23 - 24 | 3949 | 77 | |
| GenBank mRNA | AK068130.1 | - | 14 - 15 | 1820 | 77 | |
| Y12594.1 | - | 7 - 8 | 936 | 77 | ||
| Y12595.1 | - | 17 - 18 | 2205 | 77 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map