Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 7 - 8 | 1809 | 127 | |
| GenBank mRNA | AJ132280.1 | - | 7 - 8 | 1172 | 127 | |
| AK120896.1 | - | 7 - 8 | 1488 | 127 | ||
| Y12595.1 | - | 1 - 2 | 65 | 127 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map