Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 26 - 27 | 4351 | 71 | |
| GenBank mRNA | AK068130.1 | - | 17 - 18 | 2222 | 71 | |
| Y12594.1 | - | 10 - 11 | 1338 | 71 | ||
| Y12595.1 | - | 20 - 21 | 2607 | 71 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map