Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 29 - 30 | 4688 | 144 | |
| GenBank mRNA | AK068130.1 | - | 20 - 21 | 2559 | 144 | |
| Y12594.1 | - | 13 - 14 | 1675 | 144 | ||
| Y12595.1 | - | 23 - 24 | 2944 | 144 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map