Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 31 - 32 | 4998 | 78 | |
| GenBank mRNA | AK068130.1 | - | 22 - 23 | 2869 | 78 | |
| Y12594.1 | - | 15 - 16 | 1985 | 78 | ||
| Y12595.1 | - | 25 - 26 | 3254 | 78 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map