Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 9 - 10 | 2046 | 99 | |
| GenBank mRNA | AK120896.1 | - | 9 - 10 | 1725 | 99 | |
| Y12595.1 | - | 3 - 4 | 302 | 99 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map