Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 10 - 11 | 2212 | 129 | |
| GenBank mRNA | AK068130.1 | - | 1 - 2 | 83 | 129 | |
| AK120896.1 | - | 10 - 11 | 1891 | 129 | ||
| Y12595.1 | - | 4 - 5 | 468 | 129 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map