Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 11 - 12 | 2321 | 113 | |
| GenBank mRNA | AK068130.1 | - | 2 - 3 | 192 | 113 | |
| AK120896.1 | - | 11 - 12 | 2000 | 113 | ||
| Y12595.1 | - | 5 - 6 | 577 | 113 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map