Assay Details
| Gene Symbol: LOC4344164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791226.2 | XP_015646712.1 | 12 - 13 | 2535 | 81 | |
| GenBank mRNA | AK068130.1 | - | 3 - 4 | 406 | 81 | |
| AK120896.1 | - | 12 - 13 | 2214 | 81 | ||
| Y12595.1 | - | 6 - 7 | 791 | 81 | ||
More Information
Set Membership: |
|

LOC4344164
Genomic Map