Assay Details
Gene Symbol: LOC4344192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791726.1 | XP_015647212.1 | 18 - 19 | 2515 | 121 | |
GenBank mRNA | KT199367.1 | - | 18 - 19 | 2279 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
