Assay Details
| Gene Symbol: LOC4344258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789512.2 | XP_015644998.1 | 1 - 2 | 1274 | 94 | |
| GenBank mRNA | AK071934.1 | - | 1 - 2 | 1216 | 94 | |
| AK107556.1 | - | 1 - 2 | 1197 | 94 | ||
More Information
Set Membership: |
|

LOC4344258
Genomic Map