Assay Details
| Gene Symbol: LOC4344258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789512.2 | XP_015644998.1 | 2 - 3 | 1360 | 125 | |
| GenBank mRNA | AK071934.1 | - | 2 - 3 | 1302 | 125 | |
| AK107556.1 | - | 2 - 3 | 1283 | 125 | ||
More Information
Set Membership: |
|

LOC4344258
Genomic Map