Assay Details
| Gene Symbol: LOC4344427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795268.2 | XP_015650754.1 | 12 - 13 | 1706 | 105 | |
| XM_015795269.2 | XP_015650755.1 | 12 - 13 | 1606 | 105 | ||
| XM_015795271.2 | XP_015650757.1 | 13 - 14 | 1607 | 105 | ||
| GenBank mRNA | AK061938.1 | - | 5 - 6 | 549 | 105 | |
| AK067246.1 | - | 13 - 14 | 1395 | 105 | ||
More Information
Set Membership: |
|

LOC4344427
Genomic Map